Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 18, Clone: SPM510, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23441402MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23441402

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Antigen Cytokeratin 18
Immunogen Human breast cancer PMC 42 cells
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Does not react with Bovine, Canine,
Reconstitution Protein A purified
Conjugate Unlabeled
Cross Reactivity Human
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P05783
Isotype IgG1, κ
Gene ID (Entrez) 3875
Clone SPM510
Concentration 1mg/mL