Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 18, Clone: SPM510, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23441401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23441401

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Format Purified
Antigen Cytokeratin 18
Gene Accession No. P05783
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Regulatory Status RUO
Conjugate Unlabeled
Clone SPM510
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Isotype IgG1, κ
Test Specificity This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Does not react with Bovine, Canine, Hamster, Mouse, Porcine, Rat or Sheep.
Concentration 1mg/mL
Purification Method Purified
Immunogen Human breast cancer PMC 42 cells
Quantity 0.1mg
Gene ID (Entrez) 3875