Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 18, Clone: SPM265, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23435605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23435605

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Antigen Cytokeratin 18
Format Purified
Gene Accession No. P05783
Immunogen PMC-42 human breast carcinoma cells
Purification Method Purified
Quantity 0.5mL
Gene ID (Entrez) 3875
Clone SPM265
Concentration 0.2mg/mL
Formulation PBS with 0.05% BSA. Protein G purified.
Test Specificity This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. Does not react with Bovine, Canine, Hamster, Mouse, Porcine, Rat or Sheep.