Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 18, Clone: SPM265, Azide Free, Novus Biologicals™

$384.00 - $676.00

Specifications

Antigen Cytokeratin 18
Clone SPM265
Concentration 1mg/mL
Immunogen PMC-42 human breast carcinoma cells
Gene Accession No. P05783
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23440402

 
novus biologicals canada
NBP23440402MG
0.2mg Each for $676.00

NBP23440401

 
novus biologicals canada
NBP23440401MG
0.1mg Each for $384.00
Description & Specifications

Specifications

Antigen Cytokeratin 18
Clone SPM265
Concentration 1mg/mL
Immunogen PMC-42 human breast carcinoma cells
Gene Accession No. P05783
Gene ID (Entrez) 3875
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting