Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 18, Clone: SPM265, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23440402MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23440402

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Gene Accession No. P05783
Gene ID (Entrez) 3875
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.2mg
Format Purified
Clone SPM265
Primary or Secondary Primary
Conjugate Unlabeled
Isotype IgG1
Antigen Cytokeratin 18
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Concentration 1mg/mL
Reconstitution Protein A purified
Regulatory Status RUO
Purification Method Purified
Test Specificity This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. Does not react with Bovine, Canine, Hamster, Mouse, Porcine, Rat or Sheep.
Immunogen PMC-42 human breast carcinoma cells
Host Species Mouse