Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 17, Clone: SPM560, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Cytokeratin 17
Clone SPM560
Concentration 0.2mg/mL
Immunogen The cytoskeletal fraction of rat colon epithelium
Gene Accession No. Q04695
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23437801

 
novus biologicals canada
NBP23437801ML
0.1mL Each for $139.00

NBP23437805

 
novus biologicals canada
NBP23437805ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Cytokeratin 17
Clone SPM560
Concentration 0.2mg/mL
Immunogen The cytoskeletal fraction of rat colon epithelium
Gene Accession No. Q04695
Gene ID (Entrez) 3872
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rat
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting