Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 17, Clone: SPM560, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23437801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23437801

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Bovine
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rat
Host Species Mouse
Conjugate Unlabeled
Gene Accession No. Q04695
Gene ID (Entrez) 3872
Purification Method Purified
Antigen Cytokeratin 17
Immunogen The cytoskeletal fraction of rat colon epithelium
Format Purified
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Isotype IgG2b, κ
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Concentration 0.2mg/mL
Reconstitution Protein A purified
Clone SPM560
Monoclonal or Polyclonal Monoclonal