missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 17, Clone: SPM560, Azide Free, Novus Biologicals™

$396.83 - $527.00

Specifications

Antigen Cytokeratin 17
Clone SPM560
Concentration 1mg/mL
Immunogen The cytoskeletal fraction of rat colon epithelium
Gene Accession No. Q04695
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23442502 Novus Biologicals Canada
NBP23442502MG
0.2mg Each for $527.00
NBP23442501 Novus Biologicals Canada
NBP23442501MG
0.1mg Each for $396.83
Specifications Description & Specifications

Specifications

Cytokeratin 17
1mg/mL
Q04695
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM560
The cytoskeletal fraction of rat colon epithelium
3872
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot