Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 17, Clone: SPM560, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442502

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rat
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen The cytoskeletal fraction of rat colon epithelium
Quantity 0.2mg
Host Species Mouse
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3872
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Format Purified
Conjugate Unlabeled
Antigen Cytokeratin 17
Purification Method Purified
Concentration 1mg/mL
Reconstitution Protein A purified
Clone SPM560
Primary or Secondary Primary
Gene Accession No. Q04695
Isotype IgG2b, κ