Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 17, Clone: SPM560, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442501

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Isotype IgG2b, κ
Regulatory Status RUO
Primary or Secondary Primary
Cross Reactivity Bovine
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rat
Antigen Cytokeratin 17
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. Q04695
Immunogen The cytoskeletal fraction of rat colon epithelium
Purification Method Purified
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3872
Reconstitution Protein A purified
Concentration 1mg/mL
Conjugate Unlabeled
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone SPM560