Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 14, Clone: SPM263, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Cytokeratin 14
Clone SPM263
Concentration 0.2mg/mL
Immunogen A synthetic peptide of 15 amino acid from the C-terminus of human keratin 14
Gene Accession No. P02533
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23435501

 
novus biologicals canada
NBP23435501ML
0.1mL Each for $139.00

NBP23435505

 
novus biologicals canada
NBP23435505ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Cytokeratin 14
Clone SPM263
Concentration 0.2mg/mL
Immunogen A synthetic peptide of 15 amino acid from the C-terminus of human keratin 14
Gene Accession No. P02533
Gene ID (Entrez) 3861
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG3
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting