Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 14, Clone: SPM263, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23435505ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23435505

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Cytokeratin 14
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P02533
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide of 15 amino acid from the C-terminus of human keratin 14
Isotype IgG3
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3861
Reconstitution Protein A purified
Clone SPM263
Concentration 0.2mg/mL
Conjugate Unlabeled