Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 14, Clone: SPM263, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23440301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23440301

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Conjugate Unlabeled
Primary or Secondary Primary
Clone SPM263
Gene Accession No. P02533
Concentration 1mg/mL
Antigen Cytokeratin 14
Regulatory Status RUO
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Immunogen A synthetic peptide of 15 amino acid from the C-terminus of human keratin 14
Gene ID (Entrez) 3861
Host Species Mouse
Isotype IgG3
Reconstitution Protein A purified