Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 14, Clone: SPM263, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23440302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23440302

  • / Each
 

Description & Specifications

Specifications

Antigen Cytokeratin 14
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P02533
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide of 15 amino acid from the C-terminus of human keratin 14
Isotype IgG3
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3861
Clone SPM263
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Reconstitution Protein A purified
Conjugate Unlabeled