Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 14, Clone: LL002, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23427001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23427001

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 0.1mL
Formulation PBS with 0.05% BSA.
Immunogen A synthetic peptide of 15 amino acid from the C-terminus of human keratin 14
Format Purified
Isotype IgG3
Antigen Cytokeratin 14
Reconstitution Protein A purified
Purification Method Purified
Gene ID (Entrez) 3861
Concentration 0.2mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Accession No. P02533
Host Species Mouse
Conjugate Unlabeled
Regulatory Status RUO
Primary or Secondary Primary
Clone LL002