missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 14, Clone: LL002, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Cytokeratin 14
Clone LL002
Concentration 1mg/mL
Immunogen A synthetic peptide of 15 amino acid from the C-terminus of human keratin 14
Gene Accession No. P02533
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23467502 Novus Biologicals Canada
NBP23467502MG
0.2mg Each for $527.00
NBP23467501 Novus Biologicals Canada
NBP23467501MG
0.1mg Each for $384.00
Specifications Description & Specifications

Specifications

Cytokeratin 14
1mg/mL
P02533
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
LL002
A synthetic peptide of 15 amino acid from the C-terminus of human keratin 14
3861
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG3
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot