Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 14, Clone: LL002, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23467501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23467501

  • / Each
 

Description & Specifications

Specifications

Antigen Cytokeratin 14
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Gene ID (Entrez) 3861
Primary or Secondary Primary
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Isotype IgG3
Quantity 0.1mg
Host Species Mouse
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P02533
Purification Method Purified
Reconstitution Protein A purified
Clone LL002
Immunogen A synthetic peptide of 15 amino acid from the C-terminus of human keratin 14
Regulatory Status RUO