Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 10, Clone: SPM261, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Cytokeratin 10
Clone SPM261
Concentration 0.2mg/mL
Immunogen Skin extract of a human Psoriasis patient
Gene Accession No. P13645
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23296201

 
novus biologicals canada
NBP23296201ML
0.1mL Each for $139.00

NBP23296210

 
novus biologicals canada
NBP23296210ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen Cytokeratin 10
Clone SPM261
Concentration 0.2mg/mL
Immunogen Skin extract of a human Psoriasis patient
Gene Accession No. P13645
Gene ID (Entrez) 3858
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting