Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 10, Clone: SPM261, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23296210ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23296210

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Cytokeratin 10
Formulation PBS with 0.05% BSA. Protein G purified.
Host Species Mouse
Immunogen Skin extract of a human Psoriasis patient
Isotype IgG1
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Clone SPM261
Conjugate Unlabeled
Gene Accession No. P13645
Purification Method Purified
Gene ID (Entrez) 3858
Concentration 0.2mg/mL