Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 10, Clone: SPM261, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23296201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23296201

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Cytokeratin 10
Gene Accession No. P13645
Host Species Mouse
Isotype IgG1
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Clone SPM261
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Immunogen Skin extract of a human Psoriasis patient
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3858
Concentration 0.2mg/mL
Conjugate Unlabeled