Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 10, Clone: SPM261, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Cytokeratin 10
Clone SPM261
Concentration 1mg/mL
Immunogen Skin extract of a human Psoriasis patient
Gene Accession No. P13645
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23475201

 
novus biologicals canada
NBP23475201MG
0.1mg Each for $384.00

NBP23475202

 
novus biologicals canada
NBP23475202MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Cytokeratin 10
Clone SPM261
Concentration 1mg/mL
Immunogen Skin extract of a human Psoriasis patient
Gene Accession No. P13645
Gene ID (Entrez) 3858
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot