Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 10, Clone: LH2, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23466802MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23466802

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1
Quantity 0.2mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3858
Reconstitution Protein A purified
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Cytokeratin 10
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P13645
Host Species Mouse
Immunogen Skin extract of a human Psoriasis patient
Purification Method Purified
Regulatory Status RUO
Clone LH2
Concentration 1mg/mL