Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 10, Clone: LH2, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23466801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23466801

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Conjugate Unlabeled
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunogen Skin extract of a human Psoriasis patient
Clone LH2
Gene ID (Entrez) 3858
Gene Accession No. P13645
Cross Reactivity Human
Cross Reactivity Mouse
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Concentration 1mg/mL
Isotype IgG1
Quantity 0.1mg
Reconstitution Protein A purified
Purification Method Purified
Antigen Cytokeratin 10
Host Species Mouse