Please login to your online account to display your discounted pricing

Mouse anti-Cyr61/CCN1, Clone: 3A7.1B8, Novus Biologicals™

$109.00 - $309.05

Specifications

Antigen Cyr61/CCN1
Clone 3A7.1B8
Concentration 1.0mg/mL
Immunogen A synthetic peptide made to the human Cyr61/CCN1 protein sequence (between residues 140-260). [Swiss-Prot O00622]
Gene Accession No. O00622
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP236490

 
novus biologicals canada
NBP236490
0.1mg Each for $309.05

NBP236490SS

 
novus biologicals canada
NBP236490SS
0.025mg Each for $109.00
Description & Specifications

Specifications

Antigen Cyr61/CCN1
Clone 3A7.1B8
Concentration 1.0mg/mL
Immunogen A synthetic peptide made to the human Cyr61/CCN1 protein sequence (between residues 140-260). [Swiss-Prot O00622]
Gene Accession No. O00622
Gene ID (Entrez) 3491
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting