Please login to your online account to display your discounted pricing

Mouse anti-Cyclin D1, Clone: SPM587, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Cyclin D1
Clone SPM587
Concentration 0.2mg/mL
Immunogen Human recombinant full length cyclin D1 protein
Gene Accession No. P24385
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23284001

 
novus biologicals canada
NBP23284001ML
0.1mL Each for $139.00

NBP23284010

 
novus biologicals canada
NBP23284010ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen Cyclin D1
Clone SPM587
Concentration 0.2mg/mL
Immunogen Human recombinant full length cyclin D1 protein
Gene Accession No. P24385
Gene ID (Entrez) 595
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting