Please login to your online account to display your discounted pricing

Mouse anti-Cyclin D1, Clone: SPM587, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23284010ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23284010

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human recombinant full length cyclin D1 protein
Isotype IgG2a, κ
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Antigen Cyclin D1
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P24385
Purification Method Purified
Quantity 1.0mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 595
Reconstitution Protein A purified
Clone SPM587