Please login to your online account to display your discounted pricing

Mouse anti-Cyclin D1, Clone: SPM587, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Cyclin D1
Clone SPM587
Concentration 1mg/mL
Immunogen Human recombinant full length cyclin D1 protein
Gene Accession No. P24385
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23481601

 
novus biologicals canada
NBP23481601MG
0.1mg Each for $384.00

NBP23481602

 
novus biologicals canada
NBP23481602MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Cyclin D1
Clone SPM587
Concentration 1mg/mL
Immunogen Human recombinant full length cyclin D1 protein
Gene Accession No. P24385
Gene ID (Entrez) 595
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting