Please login to your online account to display your discounted pricing

Mouse anti-Cyclin D1, Clone: SPM587, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23481602MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23481602

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human recombinant full length cyclin D1 protein
Isotype IgG2a, κ
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Host Species Mouse
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity Recognizes a protein of 36kDa, identified as cyclin D1. This antibody neutralizes the activity of cyclin D1 in vivo.
Primary or Secondary Primary
Purification Method Purified
Format Purified
Gene Accession No. P24385
Quantity 0.2mg
Antigen Cyclin D1
Clone SPM587
Concentration 1mg/mL
Reconstitution Protein A purified
Gene ID (Entrez) 595