Please login to your online account to display your discounted pricing

Mouse anti-Cyclin B1, Clone: V92.1, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Cyclin B1
Clone V92.1
Concentration 0.2mg/mL
Immunogen Hamster Cyclin B1 protein
Gene Accession No. P14635
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23431201

 
novus biologicals canada
NBP23431201ML
0.1mL Each for $139.00

NBP23431205

 
novus biologicals canada
NBP23431205ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Cyclin B1
Clone V92.1
Concentration 0.2mg/mL
Immunogen Hamster Cyclin B1 protein
Gene Accession No. P14635
Gene ID (Entrez) 891
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting