Please login to your online account to display your discounted pricing

Mouse anti-Cyclin B1, Clone: V92.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23431201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23431201

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Gene Accession No. P14635
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Gene ID (Entrez) 891
Regulatory Status RUO
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 0.1mL
Concentration 0.2mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Purification Method Purified
Host Species Mouse
Formulation PBS with 0.05% BSA.
Immunogen Hamster Cyclin B1 protein
Format Purified
Reconstitution Protein A purified
Clone V92.1
Antigen Cyclin B1