Please login to your online account to display your discounted pricing

Mouse anti-Cyclin B1, Clone: V92.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23431205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23431205

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Cyclin B1
Format Purified
Formulation PBS with 0.05% BSA.
Immunogen Hamster Cyclin B1 protein
Gene ID (Entrez) 891
Conjugate Unlabeled
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Clone V92.1
Concentration 0.2mg/mL
Gene Accession No. P14635
Regulatory Status RUO
Primary or Secondary Primary