Please login to your online account to display your discounted pricing

Mouse anti-Cyclin B1, Clone: V92.1, Azide Free, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23450902MG

 View all options for this product


For Research Use Only

Catalog No. NBP23450902


Description & Specifications


Antigen Cyclin B1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Clone V92.1
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P14635
Host Species Mouse
Immunogen Hamster Cyclin B1 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 891
Reconstitution Protein A purified