Please login to your online account to display your discounted pricing

Mouse anti-Cyclin B1, Clone: V92.1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23450901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23450901

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Cyclin B1
Format Purified
Gene Accession No. P14635
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone V92.1
Concentration 1mg/mL
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Hamster Cyclin B1 protein
Regulatory Status RUO
Gene ID (Entrez) 891
Reconstitution Protein A purified
Conjugate Unlabeled