Please login to your online account to display your discounted pricing

Mouse anti-Cyclin A2, Clone: E67, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Cyclin A2
Clone E67
Concentration 0.2mg/mL
Immunogen Full length bovine Cyclin A protein
Gene Accession No. P20248
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23431101

 
novus biologicals canada
NBP23431101ML
0.1mL Each for $139.00

NBP23431105

 
novus biologicals canada
NBP23431105ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Cyclin A2
Clone E67
Concentration 0.2mg/mL
Immunogen Full length bovine Cyclin A protein
Gene Accession No. P20248
Gene ID (Entrez) 890
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mink
Cross Reactivity Monkey
Cross Reactivity Mouse
Cross Reactivity Primate
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting