Please login to your online account to display your discounted pricing

Mouse anti-CXCR1/IL-8 RA, Clone: 8F1-1-4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433390025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14333903

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Gene Accession No. P25024
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Isotype IgG2b, κ
Gene ID (Entrez) 3577
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Purification Method Affinity Purified
Antigen CXCR1/IL-8 RA
Clone 8F1-1-4
Quantity 0.025mg
Host Species Mouse
Immunogen The immunogen for this antibody was CD181.
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation PBS [pH 7.2]
Storage Requirements Store at 4°C. Do not freeze.