Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: TDM16, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23445905ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23445905

  • / Each
 

Description & Specifications

Specifications

Quantity 0.5mL
Cross Reactivity Human
Format Concentrated
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 720
Applications Immunocytochemistry
Applications Immunofluorescence
Purification Method Concentrated
Regulatory Status RUO
Formulation Culture Medium. Bioreactor concentrate.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Host Species Mouse
Clone TDM16
Conjugate Unlabeled
Gene Accession No. P0C0L4
Antigen Complement C4d
Immunogen human Complement 4d protein