Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: TDM16, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23445901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23445901

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Cross Reactivity Human
Host Species Mouse
Primary or Secondary Primary
Format Concentrated
Monoclonal or Polyclonal Monoclonal
Purification Method Concentrated
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 720
Regulatory Status RUO
Formulation Culture Medium. Bioreactor concentrate.
Antigen Complement C4d
Immunogen human Complement 4d protein
Conjugate Unlabeled
Quantity 0.1mL
Clone TDM16
Gene Accession No. P0C0L4
Isotype IgG1