Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: SPM545, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Complement C4d
Clone SPM545
Concentration 1mg/mL
Immunogen Recombinant human Complement 4d protein
Gene ID (Entrez) 721
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23442102

 
novus biologicals canada
NBP23442102MG
0.2mg Each for $527.00

NBP23442101

 
novus biologicals canada
NBP23442101MG
0.1mg Each for $384.00
Description & Specifications

Specifications

Antigen Complement C4d
Clone SPM545
Concentration 1mg/mL
Immunogen Recombinant human Complement 4d protein
Gene ID (Entrez) 721
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)