Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: SPM545, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442102

  • / Each
 

Description & Specifications

Specifications

Antigen Complement C4d
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone SPM545
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Murine
Immunogen Recombinant human Complement 4d protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 721
Test Specificity This MAb is specific to Complement 4d (C4d) and it reacts with the secreted as well as cell-bound C4d.
Reconstitution Protein A purified
Target Species Human