Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: SPM545, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442101

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Antigen Complement C4d
Immunogen Recombinant human Complement 4d protein
Regulatory Status RUO
Primary or Secondary Primary
Clone SPM545
Cross Reactivity Human
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 721
Test Specificity This MAb is specific to Complement 4d (C4d) and it reacts with the secreted as well as cell-bound C4d.
Reconstitution Protein A purified
Conjugate Unlabeled
Concentration 1mg/mL