Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: C4D205, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23446105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23446105

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Applications Immunocytochemistry
Applications Immunofluorescence
Quantity 0.5mL
Antigen Complement C4d
Formulation Culture Medium. Bioreactor concentrate.
Host Species Mouse
Immunogen Recombinant human Complement 4d protein
Isotype IgG1
Purification Method Concentrated
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 720
Clone C4D205
Cross Reactivity Human
Gene Accession No. P0C0L4
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled