Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: C4D205, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23446101ML

 View all options for this product


For Research Use Only

Catalog No. NBP23446101


Description & Specifications


Antigen Complement C4d
Applications Immunocytochemistry
Applications Immunofluorescence
Clone C4D205
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P0C0L4
Host Species Mouse
Immunogen Recombinant human Complement 4d protein
Isotype IgG1
Purification Method Concentrated
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 720