Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: C4D204, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Complement C4d
Clone C4D204
Concentration 0.2mg/mL
Immunogen Recombinant human Complement 4d protein
Gene Accession No. P0C0L4
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23423401

 
novus biologicals canada
NBP23423401ML
0.1mL Each for $139.00

NBP23423405

 
novus biologicals canada
NBP23423405ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Complement C4d
Clone C4D204
Concentration 0.2mg/mL
Immunogen Recombinant human Complement 4d protein
Gene Accession No. P0C0L4
Gene ID (Entrez) 720
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)