Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: C4D204, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23423405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23423405

  • / Each
 

Description & Specifications

Specifications

Antigen Complement C4d
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone C4D204
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P0C0L4
Host Species Mouse
Immunogen Recombinant human Complement 4d protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 720
Reconstitution Protein A purified