Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: C4D204, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23423401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23423401

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 720
Cross Reactivity Human
Clone C4D204
Regulatory Status RUO
Formulation PBS with 0.05% BSA.
Gene Accession No. P0C0L4
Purification Method Purified
Conjugate Unlabeled
Format Purified
Isotype IgG1, κ
Immunogen Recombinant human Complement 4d protein
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Concentration 0.2mg/mL
Reconstitution Protein A purified
Quantity 0.1mL
Antigen Complement C4d