missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: C4D204, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Complement C4d
Clone C4D204
Concentration 1mg/mL
Immunogen Recombinant human Complement 4d protein
Gene Accession No. P0C0L4
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23458101 Novus Biologicals Canada
NBP23458101MG
0.1mg Each for $384.00
NBP23458102 Novus Biologicals Canada
NBP23458102MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Complement C4d
1mg/mL
P0C0L4
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
C4D204
Recombinant human Complement 4d protein
720
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)