Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: C4D204, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23458101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23458101

  • / Each
 

Description & Specifications

Specifications

Format Purified
Regulatory Status RUO
Cross Reactivity Human
Conjugate Unlabeled
Quantity 0.1mg
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Isotype IgG1, κ
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Clone C4D204
Immunogen Recombinant human Complement 4d protein
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen Complement C4d
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Gene ID (Entrez) 720
Concentration 1mg/mL
Reconstitution Protein A purified
Gene Accession No. P0C0L4
Host Species Mouse