Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: C4D203, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Complement C4d
Clone C4D203
Immunogen Recombinant human Complement 4d protein
Gene Accession No. P0C0L4
Gene ID (Entrez) 720
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23446001

 
novus biologicals canada
NBP23446001ML
0.1mL Each for $139.00

NBP23446005

 
novus biologicals canada
NBP23446005ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Complement C4d
Clone C4D203
Immunogen Recombinant human Complement 4d protein
Gene Accession No. P0C0L4
Gene ID (Entrez) 720
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Concentrated
Purification Method Concentrated
Applications Immunocytochemistry
Applications Immunofluorescence