Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: C4D203, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23446005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23446005

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Host Species Mouse
Format Concentrated
Gene Accession No. P0C0L4
Immunogen Recombinant human Complement 4d protein
Regulatory Status RUO
Quantity 0.5mL
Formulation Culture Medium. Bioreactor concentrate.
Gene ID (Entrez) 720
Applications Immunocytochemistry
Applications Immunofluorescence
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Isotype IgG1
Purification Method Concentrated
Primary or Secondary Primary
Antigen Complement C4d
Clone C4D203
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.