Please login to your online account to display your discounted pricing

Mouse anti-Complement C4d, Clone: C4D203, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23446001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23446001

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Antigen Complement C4d
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P0C0L4
Host Species Mouse
Immunogen Recombinant human Complement 4d protein
Isotype IgG1
Purification Method Concentrated
Quantity 0.1mL
Primary or Secondary Primary
Gene ID (Entrez) 720
Conjugate Unlabeled
Regulatory Status RUO
Clone C4D203